Learn More
Meso Scale Discovery U-PLEX Human G-CSF ASY384-well
SDP
Supplier:  Meso Scale Discovery K251VGK2
Encompass
The Meso Scale Discovery Human G-CSF Singleplex Assay Kit quantifies G-CSF in human serum plasma and cell culture supernatants using MSD s electrochemiluminescent technology It offers high sensitivity a wide dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibody detection antibody read buffer calibrator diluent wash buffer and lyophilized calibrator It detects G-CSF concentrations from 0 3-10 000 pg/mL with high specificity and minimal cross-reactivity ideal for research applications
Catalog No. 50-000-80514
Add to Cart
Request a Quote