Learn More
Meso Scale Discovery U-PLEX Human G-CSF ASYQP
SDP
Supplier:  Meso Scale Discovery K151VGK22
Encompass
The K151VGK-1 is a Meso Scale Discovery (MSD) electrochemiluminescence immunoassay kit designed for quantitative measurement of biomarkers in biological samples This multiplex assay platform utilizes MSD s proprietary SULFO-TAG technology to deliver high sensitivity broad dynamic range and low sample volume requirements The kit includes pre-coated plates detection reagents and calibrators for streamlined workflow in research applications Compatible with MSD s MESO QuickPlex SQ 120 instrument it provides reliable reproducible results for biomarker analysis in pharmaceutical research clinical studies and academic research settings
Catalog No. 50-000-81955