Learn More
Meso Scale Discovery U-PLEX Human GITRL/TNFSF18 ASY
SDP
Supplier:  Meso Scale Discovery K151S7K2
Encompass
The Meso Scale Discovery U-PLEX Human GITRL (TNFSF18) Assay is a multiplex immunoassay kit that enables simultaneous quantification of human GITRL in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the accurate and reproducible measurement of GITRL levels which can aid in understanding immune system regulation The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and diluents valid for 50 tests
Catalog No. 50-000-78759