Learn More
Meso Scale Discovery U-PLEX Human GM-CSF ASY
SDP
Supplier:  Meso Scale Discovery K151UMK2
Encompass
The Meso Scale Discovery U-PLEX Human GM-CSF Assay Kit is a multiplex immunoassay that measures GM-CSF in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and can be fully automated on the MESO QuickPlex SQ 120 instrument The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator and controls It has a sensitivity of 0 07 pg/mL and an assay time of 3 5 hours
Catalog No. 50-000-79042