Learn More
Meso Scale Discovery U-PLEX Human HVEM/TNFRSF14 ASY
SDP
Supplier:  Meso Scale Discovery K151AGXK2
Encompass
The Meso Scale Discovery U-PLEX Human HVEM (TNFRSF14) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human Herpesvirus Entry Mediator (HVEM) in serum plasma and cell culture supernatants This assay uses MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 16 5 pg/mL This assay is for research use only and not for use in diagnostic procedures It utilizes a sandwich immunoassay format with antibodies that bind to distinct epitopes of human HVEM
Catalog No. 50-000-81904