Learn More
Meso Scale Discovery U-PLEX Human ICOS ASY
SDP
Supplier:  Meso Scale Discovery K151AHBK1
Encompass
The Meso Scale Discovery U-PLEX Human ICOS Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human ICOS in serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls It can monitor ICOS levels during immunology oncology or immunotherapy research With a short 3 5 hour protocol it delivers rapid results using as little as 12 5 L of sample
Catalog No. 50-000-80600