Learn More
Meso Scale Discovery U-PLEX Human ICOS-L/B7-H2 ASY
SDP
Supplier:  Meso Scale Discovery K151AHJK2
Encompass
The Meso Scale Discovery U-PLEX Human ICOS/L B7-H2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human Inducible T cell COStimulator (ICOS) and B7 Homolog 2 (B7-H2) biomarkers in serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and broad dynamic range The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer This assay provides reproducible and sensitive detection of human ICOS and B7-H2 with a minimal sample volume requirement of 25 L
Catalog No. 50-000-78403