Learn More
Meso Scale Discovery U-PLEX Human IFN-β Assay, Singleplex, 96-well, Sensitivity 1.2 pg/mL, Dynamic Range 1.2 - 9,530 pg/mL, Sample Types: Serum, Plasma, Cell Culture Supernatant, Research Use Only
SDP
Supplier:  Meso Scale Discovery K151ATKK2
Encompass
U-PLEX Human IFN-β Assay, Singleplex, 96-well, Sensitivity 1.2 pg/mL, Dynamic Range 1.2 - 9,530 pg/mL, Sample Types: Serum, Plasma, Cell Culture Supernatant, Research Use Only
Catalog No. NC3072156