Learn More
Meso Scale Discovery U-PLEX Human IFN2a ASYQP
SDP
Supplier:  Meso Scale Discovery K151VHK22
Encompass
The U-PLEX Human IFN2a Assay (K151VHK) is a highly sensitive electrochemiluminescence immunoassay designed to quantitatively measure human interferon alpha-2a levels in biological samples This multiplex-ready assay utilizes MSD s proprietary U-PLEX platform technology offering superior sensitivity and dynamic range compared to traditional ELISA methods The kit includes pre-coated plates detection antibodies and calibration standards enabling researchers to accurately detect IFN2a in serum plasma and cell culture supernatants for immunology and biomarker research applications
Catalog No. 50-000-78725