Learn More
Meso Scale Discovery U-PLEX Human IL-12p70 ASY
SDP
Supplier:  Meso Scale Discovery K151UAK2
Encompass
The Meso Scale Discovery U-PLEX Human IL-12p70 Assay Kit is an electrochemiluminescent immunoassay for measuring human interleukin 12 (IL-12) in serum plasma and cell culture supernatants It uses SULFO-TAG labels and multi-array technology for sensitive quantitation of IL-12p70 with detection limits from 0 74 to 1500 pg/mL The kit includes a 96-well plate detection antibodies calibrators and necessary reagents Results are analyzed on MSD s SECTOR Imager offering high sensitivity and a broad dynamic range for accurate IL-12 measurement
Catalog No. 50-000-80371