Learn More
Meso Scale Discovery U-PLEX Human IL-13 ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K151UBK4
Encompass
The Meso Scale Discovery U-PLEX Human IL-13 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-13 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager to provide a sensitive and reproducible method for measuring IL-13 with a detection range of 0 05-1000 pg/mL and an LLOQ of 0 05 pg/mL The assay can be multiplexed with other U-PLEX biomarkers and takes approximately 3 5 hours to complete This product provides a rapid ultra-sensitive immunoassay for measuring IL-13
Catalog No. 50-000-80504
Add to Cart
Request a Quote