Learn More
Meso Scale Discovery U-PLEX Human IL-16 ASY
SDP
Supplier:  Meso Scale Discovery K151USK1
Encompass
The Meso Scale Discovery U-PLEX Human IL-16 Assay Kit is a multiplex immunoassay that measures IL-16 levels in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer Diluent 2 Calibrator Blend 1 and Wash Buffer The assay takes 3 5 hours to complete and has a lower limit of detection of 0 06 pg/mL This product enables accurate quantification of IL-16 for research use only
Catalog No. 50-000-82027