Learn More
Meso Scale Discovery U-PLEX Human IL-16 ASYQP
SDP
Supplier:  Meso Scale Discovery K151USK21
Encompass
The U-PLEX Human IL-16 Assay (K151USK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-16 protein levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity detection with minimal sample volume requirements featuring a 96-well plate format for efficient processing of multiple samples The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable IL-16 quantification in research applications
Catalog No. 50-000-78246