Learn More
Meso Scale Discovery U-PLEX Human IL-17A ASY
SDP
Supplier:  Meso Scale Discovery K151UTK1
Encompass
The Meso Scale Discovery U-PLEX Human IL-17A Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-17A in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for detecting IL-17A with a lower limit of detection of 0 04 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and sample diluent The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-78247