Learn More
Meso Scale Discovery U-PLEX Human IL-17C ASY
SDP
Supplier:  Meso Scale Discovery K151WJK1
Encompass
The Meso Scale Discovery U-PLEX Human IL-17C Assay Kit is designed for the quantitative measurement of human interleukin 17C (IL-17C) in serum plasma cell culture supernatants cell and tissue lysates using an ultra-sensitive electrochemiluminescent detection technology The assay has high sensitivity and wide dynamic range to allow detection of both natural and recombinant human IL-17C This kit includes a plate coated with capture antibody detection antibody conjugated with an electrochemiluminescent label calibrator and sample diluent The assay can be run on the MESO QuickPlex SQ 120 instrument and takes approximately 3 5 hours to complete
Catalog No. 50-000-79735