Learn More
Meso Scale Discovery U-PLEX Human IL-1B ASY
SDP
Supplier:  Meso Scale Discovery K151TUK2
Encompass
The Meso Scale Discovery U-PLEX Human IL-1 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of IL-1 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and a wide dynamic range The assay has high specificity and no cross-reactivity It requires 50 L or less sample volume and can be multiplexed with other U-PLEX biomarkers The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Blend and sample diluent
Catalog No. 50-000-80367