Learn More
Meso Scale Discovery U-PLEX HUMAN IL-2 384-WELL Assay, 384-well format, Single-analyte immunoassay, Research use only, Biotin-conjugated antibodies, MSD Calibrators and Controls

Supplier: Meso Scale Discovery K251TVK2
The U-PLEX Human IL-2 384-Well Assay is a highly validated, single-analyte immunoassay designed for the quantitative measurement of human Interleukin-2 (IL-2). Utilizing the robust U-PLEX platform, this assay enables quick and efficient profiling of human IL-2 levels in various biological samples, providing essential components for high-quality data generation in a 384-well plate format. This assay is intended for research use only, ensuring researchers can obtain reliable results in their studies.
- Target analyte: Human IL-2
- Assay type: Single-analyte immunoassay
- Plate format: 384-well
- Platform: U-PLEX
- Application: Research use only
- Primary function: Measures human IL-2 levels
- Manufacturer: MESO SCALE DISCOVERY
- Quality: Validated components for high quality assays
- Multiplexing capability: Components can be combined for multiplex assays on the U-PLEX platform
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.