Learn More
Meso Scale Discovery U-PLEX Human IL-2 Assay, 5 Plates, 96-well format, dynamic range 0.70-1,900 pg/mL, LLOD 0.70 pg/mL

Supplier: Meso Scale Discovery K151TVK2
The MESO SCALE DISCOVERY K151TVK-2 U-PLEX Human IL-2 Assay is designed for the quantitative detection of human Interleukin-2 (IL-2) in a single-analyte format. This highly sensitive and robust immunoassay enables researchers to profile IL-2 levels in various biological samples, valuable for studies in cytokines, chemokines, immunology, and inflammation. The assay uses validated components that facilitate rapid and straightforward assay development, making it an effective research tool.
- Analyte: IL-2
- Species reactivity: Human
- Assay format: Single-analyte
- Plate type: 96-well
- Number of plates: 5
- Platform compatibility: U-PLEX
- Lower limit of detection: 0.70 pg/mL
- Dynamic range: 0.70-1,900 pg/mL
- Sample types: Serum, plasma, cell culture supernatant
- Detection method: Electrochemiluminescence (ECL)
- Usage statement: For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.