Learn More
Meso Scale Discovery U-PLEX Human IL-2 ASYQP
SDP
Supplier:  Meso Scale Discovery K151TVK22
Encompass
The U-PLEX Human IL-2 Assay (K151TVK) is a multiplex immunoassay platform from Meso Scale Discovery designed for quantitative measurement of interleukin-2 levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The U-PLEX format allows for flexible customizable multiplex panels that can be combined with other biomarkers Ideal for research applications in immunology oncology and drug development studies requiring precise IL-2 quantification
Catalog No. 50-000-79080