Learn More
Meso Scale Discovery U-PLEX Human IL-21 ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K151WBK4
Encompass
The Meso Scale Discovery U-PLEX Human IL-21 Assay Kit is a multiplex immunoassay that measures IL-21 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL It is for research use only
Catalog No. 50-000-78731
Add to Cart
Request a Quote