Learn More
Meso Scale Discovery U-PLEX Human IL-22 ASY
SDP
Supplier:  Meso Scale Discovery K151WIK1
Encompass
The Meso Scale Discovery U-PLEX Human IL-22 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-22 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Diluent and lyophilized U-PLEX Calibrator Blend 1 The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81476