Learn More
Meso Scale Discovery U-PLEX Human IL-3 ASY
SDP
Supplier:  Meso Scale Discovery K151XMK1
Encompass
The Meso Scale Discovery U-PLEX Human IL-3 Assay Kit is a multiplex immunoassay for measuring IL-3 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a wide dynamic range and low sample use The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG buffers diluents and calibrators It has high specificity and minimal cross-reactivity Results are analyzed with MSD software making it a valuable tool for IL-3 research
Catalog No. 50-000-80089