Learn More
Meso Scale Discovery U-PLEX Human IL-3 ASYQP
SDP
Supplier:  Meso Scale Discovery K151XMK22
Encompass
The U-PLEX Human IL-3 384-Well Assay (K251XMK) is a multiplex immunoassay platform designed for quantitative measurement of human interleukin-3 cytokine levels in biological samples This high-throughput assay utilizes 384-well plate format for efficient sample processing and features Meso Scale Discovery s electrochemiluminescence detection technology The kit provides researchers with sensitive specific detection capabilities for IL-3 biomarker analysis in clinical and research applications
Catalog No. 50-000-81792