Learn More
Meso Scale Discovery U-PLEX Human IL-31 ASY
SDP
Supplier:  Meso Scale Discovery K151WEK1
Encompass
The Meso Scale Discovery U-PLEX Human IL-31 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-31 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to MSD SULFO-TAG calibrators and an MSD Read Buffer for electrochemiluminescent signal detection on MSD instruments The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-81474