Learn More
Meso Scale Discovery U-PLEX Human IL-31 ASYQP
SDP
Supplier:  Meso Scale Discovery K151WEK22
Encompass
The U-PLEX Human IL-31 Assay (K151WEK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-31 in human biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for IL-31 detection supporting research into inflammatory and allergic conditions where IL-31 plays a key role The kit includes pre-coated plates detection antibodies and calibrators for reliable reproducible results in clinical and research applications
Catalog No. 50-000-79755