Learn More
Meso Scale Discovery U-PLEX Human IL-4 ASYQP
SDP
Supplier:  Meso Scale Discovery K151TWK21
Encompass
The U-PLEX Human IL-4 Assay (K151TWK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-4 in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable IL-4 detection in serum plasma and cell culture supernatants for research applications
Catalog No. 50-000-81989