Learn More
Meso Scale Discovery U-PLEX Human IL-5 ASY384-well
SDP
Supplier:  Meso Scale Discovery K251UOK2
Encompass
The Meso Scale Discovery U-PLEX Human IL-5 384-Well Assay Kit is a multiplex immunoassay that measures IL-5 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and assay diluent It allows simultaneous measurement of IL-5 levels along with other biomarkers in the same sample The kit has high performance and reliability for biomarker quantification
Catalog No. 50-000-80723
Add to Cart
Request a Quote