Learn More
Meso Scale Discovery U-PLEX Human IL-7 ASYQP
SDP
Supplier:  Meso Scale Discovery K151UPK21
Encompass
The U-PLEX Human IL-7 Assay (Sector K151UPK-21) is a multiplex immunoassay designed to measure interleukin-7 levels in human biological samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for IL-7 detection The assay is suitable for research applications studying immune function T-cell development and inflammatory responses providing reliable quantitative results for cytokine analysis in serum plasma and cell culture samples
Catalog No. 50-000-79724