Learn More
Meso Scale Discovery U-PLEX Human IL-8 384-Well Assay, IL-8, 384-well plate, LLOD 0.15 pg/mL, Dynamic Range 0.15-2200 pg/mL, For Research Use Only
SDP
Supplier:  Meso Scale Discovery K251TYK2
Encompass
U-PLEX Human IL-8 384-Well Assay, IL-8, 384-well plate, LLOD 0.15 pg/mL, Dynamic Range 0.15-2200 pg/mL, For Research Use Only
Catalog No. NC3199635
Add to Cart
Request a Quote