Learn More
Meso Scale Discovery U-PLEX Human IL-8 Assay, Single-analyte assay, 96-well, Lower Limit of Detection 0.15 pg/mL, Dynamic Range 0.15-2,200 pg/mL, For research use only
SDP
Supplier:  Meso Scale Discovery K151TYK2
Encompass
U-PLEX Human IL-8 Assay, Single-analyte assay, 96-well, Lower Limit of Detection 0.15 pg/mL, Dynamic Range 0.15-2,200 pg/mL, For research use only
Catalog No. NC2256031