Learn More
Meso Scale Discovery U-PLEX Human IL-23 Assay, IL-23, Human, 96-well, 5 plates, Mouse Monoclonal Antibody, 1.4 pg/mL, Dynamic Range 1.4-21,600 pg/mL, Serum, Plasma, Cell Culture Supernatant
SDP
Supplier:  Meso Scale Discovery K151WGK2
Encompass
U-PLEX Human IL-23 Assay, IL-23, Human, 96-well, 5 plates, Mouse Monoclonal Antibody, 1.4 pg/mL, Dynamic Range 1.4-21,600 pg/mL, Serum, Plasma, Cell Culture Supernatant
Catalog No. NC2944431