Learn More
Meso Scale Discovery U-PLEX Human IP-10 ASY384-well
SDP
Supplier:  Meso Scale Discovery K251UFK2
Encompass
The Meso Scale Discovery U-PLEX Human IP-10 384-Well Assay is an electrochemiluminescent immunoassay for quantifying human IP-10 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for a sensitive reproducible assay in a 384-well format Sensitivity is 0 04 pg/mL with an upper quantification limit of 10 000 pg/mL It requires 25 L of sample and takes about 3 5 hours This assay is applicable in research areas like immunology inflammation infectious diseases and oncology
Catalog No. 50-000-80700
Only null left
Add to Cart
Request a Quote