Learn More
Meso Scale Discovery U-PLEX Human MCP-1 ASYQP
SDP
Supplier:  Meso Scale Discovery K151UGK21
Encompass
The U-PLEX Human MCP-1 Assay (K151UGK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of human monocyte chemoattractant protein-1 (MCP-1/CCL2) in biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable reproducible results in research applications studying inflammation immune response and cardiovascular disease biomarkers
Catalog No. 50-000-80377