Learn More
Meso Scale Discovery U-PLEX Human MCP-2 ASY384-well
SDP
Supplier:  Meso Scale Discovery K251XHK2
Encompass
The Meso Scale Discovery U-PLEX Human MCP-2 (384-well) Assay Kit is an electrochemiluminescent immunoassay for measuring human MCP-2 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection with a range of 0 06-1500 pg/mL and a sample volume of 25 L The kit includes a 384-well plate with capture antibody detection antibody with ruthenium label calibrators and buffer enabling high-throughput MCP-2 quantification with accuracy and precision
Catalog No. 50-000-80667
Add to Cart
Request a Quote