Learn More
Meso Scale Discovery U-PLEX Human MCP-3 Assay, Single-Analyte, 96-well plate, LLOD 0.79 pg/mL, Dynamic Range 0.79 - 5000 pg/mL, Uses human serum, plasma, cell culture supernatant
SDP
Supplier:  Meso Scale Discovery K151XIK2
Encompass
The U-PLEX Human MCP-3 Assay from MESO SCALE DISCOVERY offers a robust solution for the quantitative measurement of human Monocyte Chemoattractant Protein 3 (MCP-3/CCL7). This assay is designed for research applications in immunology and inflammation, providing sensitive detection across a wide dynamic range of human biological samples.
  • High quality and ease of use with validated components
  • Enables precise quantification of human MCP-3, crucial for inflammatory response
  • Validated for diverse human sample types, including serum, plasma, and cell culture supernatant
  • Features a low limit of detection at 0.79 pg/mL
  • Wide dynamic range of 0.79 - 5,000 pg/mL for various concentrations
  • Single-analyte format optimized for dedicated MCP-3 measurement
Catalog No. NC3348166