Learn More
Meso Scale Discovery U-PLEX Human MCP-1 384-Well Assay, MCP-1, Human, 384-well, ECL detection, 0.74 pg/mL LLOD, Dynamic Range 0.74-6600 pg/mL
SDP
Supplier:  Meso Scale Discovery K251UGK2
Encompass
U-PLEX Human MCP-1 384-Well Assay, MCP-1, Human, 384-well, ECL detection, 0.74 pg/mL LLOD, Dynamic Range 0.74-6600 pg/mL
Catalog No. NC3253795
Only null left
Add to Cart
Request a Quote