Learn More
Meso Scale Discovery U-PLEX Human MDC ASY
SDP
Supplier:  Meso Scale Discovery K151UIK1
Encompass
The U-PLEX Human MDC Assay K151UIK from Meso Scale Diagnostics is a multiplex immunoassay panel that allows simultaneous quantitative measurement of 10 human inflammatory biomarkers This assay uses electrochemiluminescent detection technology and is designed for use on Meso Scale Diagnostics instrumentation The panel includes IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 and TNF- It requires 50 L of sample per well and the total assay time is approximately 3 5 hours The assay can be used for applications such as biomarker discovery translational research and clinical research
Catalog No. 50-000-79720