Learn More
Meso Scale Discovery U-PLEX Human MIF ASY
SDP
Supplier:  Meso Scale Discovery K151XJK1
Encompass
The Meso Scale Discovery U-PLEX Human MIF Assay is a multiplex immunoassay kit for measuring macrophage migration inhibitory factor (MIF) in human samples like serum plasma and more It uses electrochemiluminescence (ECL) for high sensitivity and a broad dynamic range Compatible with MSD instruments like QuickPlex it requires minimal hands-on time and quantifies MIF from pg/mL to ng/mL The kit includes a plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and adhesive seals
Catalog No. 50-000-79633