Learn More
Meso Scale Discovery U-PLEX Human MIP-3B ASY
SDP
Supplier:  Meso Scale Discovery K151VAK1
Encompass
The Meso Scale Discovery U-PLEX Human MIP-3beta Assay Kit is a multiplex immunoassay that measures human MIP-3beta in serum plasma cell culture supernatants cell and tissue lysates and other biological samples It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The assay requires 50 L or less of sample and can be read on Meso Scale Discovery MESO QuickPlex SQ 120 MESO SECTOR S 600 or MESO SECTOR S 6000 instruments The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrators
Catalog No. 50-000-81429