Learn More
Meso Scale Discovery U-PLEX Human MIP-5 ASY
SDP
Supplier:  Meso Scale Discovery K151XSK1
Encompass
The Meso Scale Discovery U-PLEX Human MIP-5 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of MIP-5 (CCL15) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance This assay allows rapid and reliable quantification of the chemokine MIP-5
Catalog No. 50-000-81794