Learn More
Meso Scale Discovery U-PLEX Human MIP-5 ASY384-well
SDP
Supplier:  Meso Scale Discovery K251XSK2
Encompass
The Meso Scale Discovery U-PLEX Human MIP-5 (384-well) Assay Kit is a multiplex immunoassay that measures human MIP-5 protein levels across two 384-well plates using electrochemiluminescence detection technology This assay kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions used to generate a standard curve It provides high sensitivity broad dynamic range and requires low sample volume This assay kit can be used for biomarker research and drug discovery applications
Catalog No. 50-000-80691
Only null left
Add to Cart
Request a Quote