Learn More
Meso Scale Discovery U-PLEX Human MMP-2 ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K151AHKK4
Encompass
The Meso Scale Discovery U-PLEX Human MMP-2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of MMP-2 in human serum plasma urine saliva CSF and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range low sample consumption and rapid time to results The assay requires 50 L or less sample volume and can be read on MSD instrument platforms It includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure MMP-2 concentrations
Catalog No. 50-000-81652
Add to Cart
Request a Quote