Learn More
Meso Scale Discovery U-PLEX Human MMP-2 ASYQP
SDP
Supplier:  Meso Scale Discovery K151AHKK21
Encompass
The Meso Scale Discovery U-PLEX Human MMP-2 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of MMP-2 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of MMP-2 levels The assay has a detection range of 0 03-400 ng/mL and exhibits no cross-reactivity with other MMP family members It requires 50 L sample volume and results can be read within 3 5 hours using an MSD instrument The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and assay diluent
Catalog No. 50-000-81588