Learn More
Meso Scale Discovery U-PLEX Human MMP-7 ASY
SDP
Supplier:  Meso Scale Discovery K151AHEK1
Encompass
The Meso Scale Discovery U-PLEX Human MMP-7 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of MMP-7 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of MMP-7 levels down to 0 04 ng/mL The assay can be fully automated on MSD s SECTOR Imager platform and has a total assay time of 3 5 hours for 96 tests The assay kit includes a 96-well plate coated with capture antibody detection antibody labeled with MSD SULFO-TAG calibrators and assay diluent The assay has high specificity for active MMP-7 with no cross-reactivity to other MMPs
Catalog No. 50-000-78808