Learn More
Meso Scale Discovery U-PLEX Human MMP-7 ASYQP
SDP
Supplier:  Meso Scale Discovery K151AHEK21
Encompass
The Meso Scale Discovery U-PLEX Human MMP-7 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of MMP-7 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and rapid detection of MMP-7 levels down to 0 04 ng/mL The assay requires a small sample volume of 50 L and has a 3 5 hour total assay time The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrator solutions and sample diluent It provides a simple reproducible method to quantify MMP-7 levels for life science research applications
Catalog No. 50-000-81656