Learn More
Meso Scale Discovery U-PLEX Human PlGF ASYQP
SDP
Supplier:  Meso Scale Discovery K151B8K21
Encompass
The Meso Scale Discovery U-PLEX Human PD-L2 Assay is a multiplex immunoassay kit for measuring human PD-L2 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls This assay enables rapid reproducible quantification of PD-L2 levels crucial for immunotherapy research
Catalog No. 50-000-81975