Learn More
Meso Scale Discovery U-PLEX Human proMMP-9 ASYQP
SDP
Supplier:  Meso Scale Discovery K151AHLK22
Encompass
The Meso Scale Discovery U-PLEX Human ProMMP-9 Assay is a multiplex immunoassay that measures ProMMP-9 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can be fully automated on the MSD instrument platforms The U-PLEX format allows simultaneous measurement of ProMMP-9 with other biomarkers in human samples Overall this is a highly sensitive and reproducible assay for quantifying ProMMP-9 levels in various human sample types
Catalog No. 50-000-81581