Learn More
Meso Scale Discovery U-PLEX Human proMMP-9 ASYQP
SDP
Supplier:  Meso Scale Discovery K151AHLK21
Encompass
The Meso Scale Discovery U-PLEX Human ProMMP-9 Assay is a multiplex immunoassay for measuring ProMMP-9 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity broad dynamic range and precision Requiring 50 L or less sample volume it can be automated on MSD platforms The U-PLEX format allows simultaneous measurement with other biomarkers This assay enables accurate quantitation of ProMMP-9 aiding research in diseases like cancer arthritis cardiovascular and neurological disorders
Catalog No. 50-000-81590