Learn More
Meso Scale Discovery U-PLEX Human PYY total ASYQP
SDP
Supplier:  Meso Scale Discovery K1516BK22
Encompass
The MSD U-PLEX Total PYY Assay is a human-specific immunoassay designed to measure total peptide YY (PYY) levels in biological samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for PYY quantification The assay is suitable for research applications studying metabolic regulation appetite control and gastrointestinal hormone function providing researchers with a reliable tool for PYY biomarker analysis
Catalog No. 50-000-78829