Learn More
Meso Scale Discovery U-PLEX Human RANTES ASYQP
SDP
Supplier:  Meso Scale Discovery K151A2K21
Encompass
The Meso Scale Discovery U-PLEX Human RANTES Assay is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of RANTES (CCL5) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and buffers and reagents required to perform the assay It can accurately quantify RANTES concentrations between 0 06 - 2500 pg/mL with minimal hands-on time using an MSD instrument
Catalog No. 50-000-79538